missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(4-Methylsulfonylbenzoyl)propionic acid, 98%

Manufacturer:  Alfa Aesar™ L1275003

 View more versions of this product

Catalog No. AAL1275003



Specifications

Specifications

3-(4-Methylsulfonylbenzoyl)propionic acid
1g
C11H12O5S
2134110
LOIRMIGGBFNYOZ-UHFFFAOYSA-N
4-(4-methylsulfonylphenyl)-4-oxobutanoic acid
603015
98%
168°C to 172°C
7028-79-7
MFCD00082738
3-4-methylsulfonylbenzoyl propionic acid, 4-4-methylsulfonyl phenyl-4-oxobutanoic acid, 4-4-methanesulfonylphenyl-4-oxobutanoic acid, 4-4-methylsulfonyl phenyl-4-oxobutanoic acid, 3-4-methylsulphonylbenzoyl propionic acid, 3-4-methanesulfonylbenzoyl propionic acid, 4-4-methylsulfonylphenyl-4-oxobutanoic acid, 4-4-methylsulfonylphenyl-4-oxo-butanoic acid, 3-4-methylsulfonylbenzoyl propanoic acid, 4-4-methanesulfonyl phenyl-4-oxobutanoic acid
CS(=O)(=O)C1=CC=C(C=C1)C(=O)CCC(=O)O
256.272
256.28

Chemical Identifiers

7028-79-7
256.272
LOIRMIGGBFNYOZ-UHFFFAOYSA-N
603015
CS(=O)(=O)C1=CC=C(C=C1)C(=O)CCC(=O)O
C11H12O5S
MFCD00082738
3-4-methylsulfonylbenzoyl propionic acid, 4-4-methylsulfonyl phenyl-4-oxobutanoic acid, 4-4-methanesulfonylphenyl-4-oxobutanoic acid, 4-4-methylsulfonyl phenyl-4-oxobutanoic acid, 3-4-methylsulphonylbenzoyl propionic acid, 3-4-methanesulfonylbenzoyl propionic acid, 4-4-methylsulfonylphenyl-4-oxobutanoic acid, 4-4-methylsulfonylphenyl-4-oxo-butanoic acid, 3-4-methylsulfonylbenzoyl propanoic acid, 4-4-methanesulfonyl phenyl-4-oxobutanoic acid
4-(4-methylsulfonylphenyl)-4-oxobutanoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents