Please login to your online account to display your discounted pricing

3-[(4-Methylpiperazin-1-yl)methyl]benzaldehyde, ≥95%, Maybridge


Quantity 5g

Manufacturer: fisher scientific company  CC45304DE

 View more versions of this product

Catalog No. CC45304DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 218.3
Quantity 5g
Assay 95%
CAS Max % 100.0
Chemical Name or Material 3-[(4-Methylpiperazin-1-yl)methyl]benzaldehyde
MDL Number MFCD08271925
CAS Min % 94.5
Health Hazard 1 GHS Signal Word: Danger
Molecular Formula C13H18N2O
CAS 859850-88-7
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.
Assay Percent Range 95%

Specifications

Chemical Name or Material 3-[(4-Methylpiperazin-1-yl)methyl]benzaldehyde
CAS 859850-88-7
CAS Min % 94.5
CAS Max % 100.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C13H18N2O
Formula Weight 218.3
MDL Number MFCD08271925
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.