Please login to your online account to display your discounted pricing

3-(4-Methylpiperazin-1-yl)aniline, 97%, Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC35914DE

 View more versions of this product

Catalog No. CC35914DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 191.28
Quantity 5g
Assay 97%
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C11H17N3
Chemical Name or Material 3-(4-Methylpiperazin-1-yl)aniline
CAS 148546-99-0

Specifications

Chemical Name or Material 3-(4-Methylpiperazin-1-yl)aniline
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H17N3
Formula Weight 191.28