Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(4-Methylphenyl)propionic acid, 98+%

$97.49 - $367.65

Chemical Identifiers

Chemical Name or Material 3-(4-Methylphenyl)propionic acid
CAS 1505-50-6
Molecular Formula C10H12O2
Formula Weight 164.2
MDL Number MFCD00016561
Beilstein 2046164
Synonym 4-Methylhydrocinnamic acid; 3-(p-Tolyl)propionic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1451306

  SDS
alfa aesar™
A1451306
5g Each for $97.49

AAA1451309

  SDS
alfa aesar™
A1451309
10g Each for $171.77

AAA1451314

  SDS
alfa aesar™
A1451314
25g Each for $367.65
Description & Specifications

Specifications

Formula Weight 164.2
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Chemical Name or Material 3-(4-Methylphenyl)propionic acid
CAS 1505-50-6
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Beilstein 2046164
Molecular Formula C10H12O2
MDL Number MFCD00016561
Synonym 4-Methylhydrocinnamic acid; 3-(p-Tolyl)propionic acid
Melting Point 115° to 117°C
Quantity 5g
Assay ≥98%

Applications
3-(p-Tolyl)propionic acid was used during the coupling of 1-(3-dimethylaminopropyl)-3-ethylcarbodiimide with 4,5-diphenyl-2-(2-p-tolyl-ethyl)-4,5-dihydro-1H-imidazole resin.

Solubility
Soluble in methanol.

Notes
Keep container tightly closed. Store in a cool and dark place. Store away from incompatible materials such as oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.