missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(4-Methylphenyl)propionic acid, 98+%

Manufacturer:  Alfa Aesar™ A1451314

 View more versions of this product

Catalog No. AAA1451314



Description

Description

Applications
3-(p-Tolyl)propionic acid was used during the coupling of 1-(3-dimethylaminopropyl)-3-ethylcarbodiimide with 4,5-diphenyl-2-(2-p-tolyl-ethyl)-4,5-dihydro-1H-imidazole resin.

Solubility
Soluble in methanol.

Notes
Keep container tightly closed. Store in a cool and dark place. Store away from incompatible materials such as oxidizing agents.
Specifications

Specifications

3-(4-Methylphenyl)propionic acid
25g
C10H12O2
2046164
LDYGRLNSOKABMM-UHFFFAOYSA-N
3-(4-methylphenyl)propanoic acid
73927
≥98%
115°C to 117°C
1505-50-6
MFCD00016561
3-4-methylphenyl propionic acid, 3-p-tolyl propionic acid, 3-p-tolyl propanoic acid, benzenepropanoic acid, 4-methyl, 3-4-methylphenyl propanoic acid, 4-methylphenylpropionic acid, 3-p-tolylpropanoic acid, p-methylhydrocinnamic acid, 4-methyl phenyl propionic acid, 4-methylhydrocinnamic acid
CC1=CC=C(C=C1)CCC(=O)O
164.204
164.2

Chemical Identifiers

1505-50-6
164.204
LDYGRLNSOKABMM-UHFFFAOYSA-N
73927
CC1=CC=C(C=C1)CCC(=O)O
C10H12O2
MFCD00016561
3-4-methylphenyl propionic acid, 3-p-tolyl propionic acid, 3-p-tolyl propanoic acid, benzenepropanoic acid, 4-methyl, 3-4-methylphenyl propanoic acid, 4-methylphenylpropionic acid, 3-p-tolylpropanoic acid, p-methylhydrocinnamic acid, 4-methyl phenyl propionic acid, 4-methylhydrocinnamic acid
3-(4-methylphenyl)propanoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents