missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(4-Methylphenyl)-5-phenyl-1,2,4-oxadiazole, 97%

Manufacturer:  Alfa Aesar™ H54813MD

 View more versions of this product

Catalog No. AAH54813MD



Specifications

Specifications

3-(4-Methylphenyl)-5-phenyl-1,2,4-oxadiazole
16151-03-4
MFCD02650635
GZLTUHPTDPXMHI-UHFFFAOYSA-N
3-(4-methylphenyl)-5-phenyl-1,2,4-oxadiazole
583987
97%
250mg
C15H12N2O
1,2,4-oxadiazole, 3-4-methylphenyl-5-phenyl
CC1=CC=C(C=C1)C2=NOC(=N2)C3=CC=CC=C3
236.274
236.27

Chemical Identifiers

16151-03-4
236.274
GZLTUHPTDPXMHI-UHFFFAOYSA-N
583987
CC1=CC=C(C=C1)C2=NOC(=N2)C3=CC=CC=C3
C15H12N2O
MFCD02650635
1,2,4-oxadiazole, 3-4-methylphenyl-5-phenyl
3-(4-methylphenyl)-5-phenyl-1,2,4-oxadiazole
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents