Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(4-Methylphenyl)-1H-pyrazole-4-carboxaldehyde, 97%

$125.20 - $545.45

Chemical Identifiers

Chemical Name or Material 3-(4-Methylphenyl)-1H-pyrazole-4-carboxaldehyde
CAS 350988-62-4
Molecular Formula C11H10N2O
Formula Weight 186.22
MDL Number MFCD01133694
Synonym 3-(p-Tolyl)-1H-pyrazole-4-carboxaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5045906

  SDS
alfa aesar™
H5045906
5g Each for $545.45

AAH5045903

  SDS
alfa aesar™
H5045903
1g Each for $125.20
Description & Specifications

Specifications

Formula Weight 186.22
Chemical Name or Material 3-(4-Methylphenyl)-1H-pyrazole-4-carboxaldehyde
Assay 97%
Molecular Formula C11H10N2O
MDL Number MFCD01133694
CAS 350988-62-4
Synonym 3-(p-Tolyl)-1H-pyrazole-4-carboxaldehyde
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.