Please login to your online account to display your discounted pricing

3,4-Methylenedioxyphenylboronic acid, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC359580050

 View more versions of this product

Catalog No. AC359580050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay 97.5 to 106% (Acid)
Formula Weight 165.94
Physical Form Powder
Quantity 5g
Color White to off-white
CAS Max % 100.0
Chemical Name or Material 3, 4-Methylenedioxyphenylboronic acid
Molecular Formula C7H7BO4
Assay Percent Range 98%
CAS 94839-07-3
CAS Min % 97.5
MDL Number MFCD01009695
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 3, 4-Methylenedioxyphenylboronic acid
CAS 94839-07-3
CAS Min % 97.5
CAS Max % 100.0
Assay 97.5 to 106% (Acid)
Assay Percent Range 98%
Molecular Formula C7H7BO4
Formula Weight 165.94
MDL Number MFCD01009695
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents