missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Methylenedioxynitrobenzene 98.0+%, TCI America™

$201.67 - $1,695.67

Chemical Identifiers

CAS 2620-44-2
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00005824
InChI Key SNWQAKNKGGOVMO-UHFFFAOYSA-N
Synonym 1,2-methylenedioxy-4-nitrobenzene, 1,3-benzodioxole, 5-nitro, 3,4-methylenedioxy-1-nitrobenzene, 3,4-methylenedioxynitrobenzene, 5-nitro-2h-1,3-benzodioxole, 5-nitrobenzo d 1,3 dioxole, 5-nitrobenzodioxole, benzene, 1,2-methylenedioxy-4-nitro, methylenedioxynitrobenzene
PubChem CID 75798
IUPAC Name 5-nitro-1,3-benzodioxole
SMILES C1OC2=C(O1)C=C(C=C2)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M084825G View Documents Tci America
M084825G
25g Each for $201.67
 
M0848500G View Documents Tci America
M0848500G
500g Each for $1,695.67
 
Specifications

Chemical Identifiers

2620-44-2
167.12
SNWQAKNKGGOVMO-UHFFFAOYSA-N
75798
C1OC2=C(O1)C=C(C=C2)[N+](=O)[O-]
C7H5NO4
MFCD00005824
1,2-methylenedioxy-4-nitrobenzene, 1,3-benzodioxole, 5-nitro, 3,4-methylenedioxy-1-nitrobenzene, 3,4-methylenedioxynitrobenzene, 5-nitro-2h-1,3-benzodioxole, 5-nitrobenzo d 1,3 dioxole, 5-nitrobenzodioxole, benzene, 1,2-methylenedioxy-4-nitro, methylenedioxynitrobenzene
5-nitro-1,3-benzodioxole

Specifications

Yellow
25g
C7H5NO4
1,2-methylenedioxy-4-nitrobenzene, 1,3-benzodioxole, 5-nitro, 3,4-methylenedioxy-1-nitrobenzene, 3,4-methylenedioxynitrobenzene, 5-nitro-2h-1,3-benzodioxole, 5-nitrobenzo d 1,3 dioxole, 5-nitrobenzodioxole, benzene, 1,2-methylenedioxy-4-nitro, methylenedioxynitrobenzene
C1OC2=C(O1)C=C(C=C2)[N+](=O)[O-]
167.12
167.12
≥98.0% (GC)
148°C
2620-44-2
MFCD00005824
SNWQAKNKGGOVMO-UHFFFAOYSA-N
5-nitro-1,3-benzodioxole
75798
Crystal-Powder at 20°C
3,4-Methylenedioxynitrobenzene
Safety and Handling

Safety and Handling

TSCA : Yes