missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Methylenedioxyaniline, TCI America™

Manufacturer:  TCI America M0741100G

 View more versions of this product

Catalog No. M0741100G



Specifications

Specifications

3,4-Methylenedioxyaniline
Undesignated
100g
C7H7NO2
3,4-methylenedioxy aniline, benzo d 1,3 dioxol-5-amine, 5-amino-1,3-benzodioxole, 3,4-methylenedioxyaniline, 4-amino-1,2-methylenedioxybenzene, 5-aminobenzodioxole, methylenedioxyaniline, 2h-1,3-benzodioxol-5-amine, 1-amino-3,4-methylenedioxybenzene, 1-amino-3,4-dimethylenedioxybenzene
C1OC2=C(O1)C=C(C=C2)N
137.138
137.14
≥98.0% (GC,T)
156°C
44°C
14268-66-7
MFCD00005832
XGNXYCFREOZBOL-UHFFFAOYSA-N
1,3-benzodioxol-5-amine
84310
Crystal-Powder at 20°C

Chemical Identifiers

14268-66-7
137.138
XGNXYCFREOZBOL-UHFFFAOYSA-N
84310
C1OC2=C(O1)C=C(C=C2)N
C7H7NO2
MFCD00005832
3,4-methylenedioxy aniline, benzo d 1,3 dioxol-5-amine, 5-amino-1,3-benzodioxole, 3,4-methylenedioxyaniline, 4-amino-1,2-methylenedioxybenzene, 5-aminobenzodioxole, methylenedioxyaniline, 2h-1,3-benzodioxol-5-amine, 1-amino-3,4-methylenedioxybenzene, 1-amino-3,4-dimethylenedioxybenzene
1,3-benzodioxol-5-amine
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Refrigerator

Documents