missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-(Methylenedioxy)thiobenzamide, 97%

$68.96 - $230.89

Chemical Identifiers

CAS 15884-65-8
Molecular Formula C8H7NO2S
Formula Weight 181.21
MDL Number MFCD02677739
Synonym 1,3-Benzodioxole-5-thiocarboxamide
Chemical Name or Material 3,4-(Methylenedioxy)thiobenzamide
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5183503 SDS Alfa Aesar™
H5183503
1g Each for $230.89
AAH51835MD SDS Alfa Aesar™
H51835MD
250mg Each for $68.96
Specifications Description & Specifications

Chemical Identifiers

15884-65-8
181.21
1,3-Benzodioxole-5-thiocarboxamide
C8H7NO2S
MFCD02677739
3,4-(Methylenedioxy)thiobenzamide

Specifications

15884-65-8
181.21
1,3-Benzodioxole-5-thiocarboxamide
3,4-(Methylenedioxy)thiobenzamide
C8H7NO2S
MFCD02677739
1g
97%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning