Please login to your online account to display your discounted pricing

Alfa Aesar™ 3',4'-(Methylenedioxy)propiophenone, 98%

$151.16 - $623.41

Chemical Identifiers

Chemical Name or Material 3',4'-(Methylenedioxy)propiophenone
CAS 28281-49-4
Molecular Formula C10H10O3
Formula Weight 178.19
MDL Number MFCD00016652
Beilstein 165729
Synonym 1-(1,3-Benzodioxol-5-yl)-1-propanone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1446506

  SDS
alfa aesar™
A1446506
5g Each for $151.16

AAA1446514

  SDS
alfa aesar™
A1446514
25g Each for $623.41
Description & Specifications

Specifications

Formula Weight 178.19
Chemical Name or Material 3',4'-(Methylenedioxy)propiophenone
Assay 98%
Molecular Formula C10H10O3
MDL Number MFCD00016652
CAS 28281-49-4
Beilstein 165729
Synonym 1-(1,3-Benzodioxol-5-yl)-1-propanone
Melting Point 35° to 39°C
Boiling Point 165° to 168°C (20mmHg)
Quantity 5g
RTECS Number UC0440600