Please login to your online account to display your discounted pricing

3,4-(Methylenedioxy)phenylacetonitrile, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3, 4-(Methylenedioxy)phenylacetonitrile
Name Note 98%
CAS 4439-02-5
CAS Min % 97.5
CAS Max % 100.0
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C9H7NO2
Formula Weight 161.16g/mol
MDL Number MFCD00005835
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC163350100

  SDS
fisher scientific acros
163350100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Color Pale yellow
Quantity 10g
Chemical Name or Material 3, 4-(Methylenedioxy)phenylacetonitrile
Name Note 98%
CAS 4439-02-5
CAS Min % 97.5
CAS Max % 100.0
Physical Form Pale yellow low melting solid
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C9H7NO2
Formula Weight 161.16g/mol
MDL Number MFCD00005835
Synonym 1, 3-Benzodioxole-5-acetonitrile
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.