missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-(Methylenedioxy)cinnamic acid, predominantly trans, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1012236

 View more versions of this product

Catalog No. AAA1012236


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3,4- (Methylenedioxy)cinnamic acid
2373-80-0
C10H8O4
500 g
3,4-methylenedioxycinnamic acid, 3,4-methylenedioxy cinnamic acid, 3-benzo d 1,3 dioxol-5-yl acrylic acid, unii-x8h0wpj08y, 3-1,3-benzodioxol-5-yl acrylic acid, 2e-3-2h-1,3-benzodioxol-5-yl prop-2-enoic acid, methylenedioxycinnamic acid, x8h0wpj08y, 2e-3-1,3-benzodioxol-5-yl acrylic acid, 2e-3-1,3-benzodioxol-5-yl-2-propenoic acid
C1OC2=C(O1)C=C(C=C2)C=CC(=O)O
192.17
CHEBI:81482
0.99
predominantly trans
∼243°C (decomposition)
MFCD00005837
84599
QFQYZMGOKIROEC-DUXPYHPUSA-N
(E)-3-(1,3-benzodioxol-5-yl)prop-2-enoic acid
643181
192.17

Chemical Identifiers

2373-80-0
192.17
QFQYZMGOKIROEC-DUXPYHPUSA-N
643181
(E)-3-(1,3-benzodioxol-5-yl)prop-2-enoic acid
C10H8O4
MFCD00005837
3,4-methylenedioxycinnamic acid, 3,4-methylenedioxy cinnamic acid, 3-benzo d 1,3 dioxol-5-yl acrylic acid, unii-x8h0wpj08y, 3-1,3-benzodioxol-5-yl acrylic acid, 2e-3-2h-1,3-benzodioxol-5-yl prop-2-enoic acid, methylenedioxycinnamic acid, x8h0wpj08y, 2e-3-1,3-benzodioxol-5-yl acrylic acid, 2e-3-1,3-benzodioxol-5-yl-2-propenoic acid
CHEBI:81482
C1OC2=C(O1)C=C(C=C2)C=CC(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

EINECSNumber : 219-151-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only