Please login to your online account to display your discounted pricing

3,4-(Methylenedioxy)cinnamic acid, 99%, predominantly trans, ACROS Organics™


Quantity 100g

Manufacturer: thermo fisher scientific  163891000

 View more versions of this product

Catalog No. AC163891000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 192.17
Assay ≥98.5%
Physical Form Granular Powder
Quantity 100g
Color White to light yellow
Melting Point 242° to 244°C
EINECS Number 219-151-0
TSCA TSCA
Beilstein 19, 278
CAS Max % 100.0
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
CAS Min % 98.5
Molecular Formula C10H8O4
MDL Number MFCD00005837
CAS 2373-80-0
Name Note 99%
Chemical Name or Material 3, 4-(Methylenedioxy)cinnamic acid
Assay Percent Range 99%

Specifications

Chemical Name or Material 3, 4-(Methylenedioxy)cinnamic acid
Name Note 99%
CAS 2373-80-0
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10H8O4
Formula Weight 192.17
MDL Number MFCD00005837
Accessories

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.

SDS & Safety Documents