Please login to your online account to display your discounted pricing

3,4-(Methylenedioxy)cinnamic acid, 99%, predominantly trans, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3, 4-(Methylenedioxy)cinnamic acid
Name Note 99%
CAS 2373-80-0
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10H8O4
Formula Weight 192.17
MDL Number MFCD00005837
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC163891000

  SDS
thermo fisher scientific
163891000
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 192.17
Physical Form Granular Powder
Assay ≥98.5%
Quantity 100g
Color White to light yellow
Melting Point 242° to 244°C
EINECS Number 219-151-0
TSCA TSCA
Beilstein 19, 278
CAS Max % 100.0
CAS Min % 98.5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Chemical Name or Material 3, 4-(Methylenedioxy)cinnamic acid
Name Note 99%
Assay Percent Range 99%
Molecular Formula C10H8O4
MDL Number MFCD00005837
CAS 2373-80-0
Safety and Handling

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.