missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Methylenedioxy-beta-nitrostyrene, 98%

$102.61 - $394.92

Chemical Identifiers

CAS 1485-00-3
Molecular Formula C9H7NO4
Formula Weight 193.16
MDL Number MFCD00014575
Beilstein 192350
Synonym 1-(3,4-Methylenedioxy)phenyl-2-nitroethene; MNS
Chemical Name or Material 3,4-Methylenedioxy-beta-nitrostyrene
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0913918 SDS Alfa Aesar™
L0913918
50g Each for $394.92
AAL0913909 SDS Alfa Aesar™
L0913909
10g Each for $102.61
Specifications Description & Specifications

Chemical Identifiers

1485-00-3
193.16
192350
3,4-Methylenedioxy-beta-nitrostyrene
C9H7NO4
MFCD00014575
1-(3,4-Methylenedioxy)phenyl-2-nitroethene; MNS

Specifications

1485-00-3
193.16
192350
159° to 163°C
3,4-Methylenedioxy-beta-nitrostyrene
C9H7NO4
MFCD00014575
1-(3,4-Methylenedioxy)phenyl-2-nitroethene; MNS
50g
98%

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : WL5270000