Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-(Methylenedioxy)benzylamine, 97%

Chemical Identifiers

Chemical Name or Material 3,4-(Methylenedioxy)benzylamine
CAS 2620-50-0
Molecular Formula C8H9NO2
Formula Weight 151.17
MDL Number MFCD00005840
UN Number UN2735
Synonym (1,3-Benzodioxol-5-yl)methylamine; Piperonylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2183706

  SDS
alfa aesar™
B2183706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2183714

  SDS
alfa aesar™
B2183714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2183722

  SDS
alfa aesar™
B2183722
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 151.17
Health Hazard 2 GHS H Statement
H314
Causes severe skin burns and eye damage.
Assay 97%
Quantity 5g
UN Number UN2735
CAS 2620-50-0
Boiling Point 136° to 137°C (11mmHg)
Odor Amine-like
Chemical Name or Material 3,4-(Methylenedioxy)benzylamine
MDL Number MFCD00005840
Molecular Formula C8H9NO2
Health Hazard 1 Danger
Synonym (1,3-Benzodioxol-5-yl)methylamine; Piperonylamine
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point >110°C (230°F)

Applications
Piperonylamine is a metabolite of dopamine analogs. It is a pharmaceutical intermediate.

Solubility
Sparingly soluble in water. (0.9 g/100 mL).

Notes
Air sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store at 2-8°C.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314
Causes severe skin burns and eye damage.