Learn More
3,4-(Methylenedioxy)benzyl isothiocyanate, 94%, Thermo Scientific™
$103.19 - $103.19
Chemical Identifiers
CAS | 4430-47-1 |
---|---|
Molecular Formula | C9H7NO2S |
Molecular Weight (g/mol) | 193.22 |
MDL Number | MFCD00041217 |
InChI Key | PUJWRDBPAFJUJW-UHFFFAOYSA-N |
Synonym | 3,4-methylenedioxy benzyl isothiocyanate, 1,3-benzodioxol-5-ylmethyl isothiocyanate, 5-isothiocyanatomethyl-1,3-benzodioxole, 3,4-methylenedioxybenzyl isothiocyanate, 5-isothiocyanatomethyl-2h-1,3-benzodioxole, 5-isothiocyanatomethyl-benzo 1,3 dioxole, 5-isothiocyanatomethyl benzo d 1,3 dioxole, acmc-20angn, pubchem9883, akos bbr-002798 |
PubChem CID | 2795371 |
IUPAC Name | 5-(isothiocyanatomethyl)-1,3-benzodioxole |
SMILES | S=C=NCC1=CC=C2OCOC2=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1050103
|
Thermo Scientific Chemicals
L1050103 |
1 g |
Each for $103.19
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications3,4-(Methylenedioxy)benzyl isothiocyanate is used as intermediates. Used in nucleophilic addition reactions.
Solubility
Hydrolyses in water.
Notes
Moisture Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from moisture, alocohol, amines, water.
Chemical Identifiers
4430-47-1 | |
193.22 | |
PUJWRDBPAFJUJW-UHFFFAOYSA-N | |
2795371 | |
S=C=NCC1=CC=C2OCOC2=C1 |
C9H7NO2S | |
MFCD00041217 | |
3,4-methylenedioxy benzyl isothiocyanate, 1,3-benzodioxol-5-ylmethyl isothiocyanate, 5-isothiocyanatomethyl-1,3-benzodioxole, 3,4-methylenedioxybenzyl isothiocyanate, 5-isothiocyanatomethyl-2h-1,3-benzodioxole, 5-isothiocyanatomethyl-benzo 1,3 dioxole, 5-isothiocyanatomethyl benzo d 1,3 dioxole, acmc-20angn, pubchem9883, akos bbr-002798 | |
5-(isothiocyanatomethyl)-1,3-benzodioxole |
Specifications
4430-47-1 | |
139°C to 140°C (1 mmHg) | |
C9H7NO2S | |
MFCD00041217 | |
UN2922 | |
Moisture sensitive | |
Hydrolyses in water. | |
S=C=NCC1=CC=C2OCOC2=C1 | |
193.22 | |
193.23 | |
3,4-(Methylenedioxy)benzyl isothiocyanate |
1.32 | |
Pungent | |
1.615 | |
1 g | |
1428941 | |
3,4-methylenedioxy benzyl isothiocyanate, 1,3-benzodioxol-5-ylmethyl isothiocyanate, 5-isothiocyanatomethyl-1,3-benzodioxole, 3,4-methylenedioxybenzyl isothiocyanate, 5-isothiocyanatomethyl-2h-1,3-benzodioxole, 5-isothiocyanatomethyl-benzo 1,3 dioxole, 5-isothiocyanatomethyl benzo d 1,3 dioxole, acmc-20angn, pubchem9883, akos bbr-002798 | |
PUJWRDBPAFJUJW-UHFFFAOYSA-N | |
5-(isothiocyanatomethyl)-1,3-benzodioxole | |
2795371 | |
94% |
Safety and Handling
GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H312-H314-H331-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures