missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-(Methylenedioxy)benzyl isothiocyanate, 94%

Manufacturer:  Alfa Aesar™ L1050106

 View more versions of this product

Catalog No. AAL1050106



Description

Description

Applications
3,4-(Methylenedioxy)benzyl isothiocyanate is used as intermediates. Used in nucleophilic addition reactions.

Solubility
Hydrolyses in water.

Notes
Moisture Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from moisture, alocohol, amines, water.
Specifications

Specifications

3,4-(Methylenedioxy)benzyl isothiocyanate
139°C to 140°C (1mmHg)
4430-47-1
MFCD00041217
1428941
PUJWRDBPAFJUJW-UHFFFAOYSA-N
5-(isothiocyanatomethyl)-1,3-benzodioxole
2795371
94%
Pungent
5g
C9H7NO2S
UN2922
3,4-methylenedioxy benzyl isothiocyanate, 1,3-benzodioxol-5-ylmethyl isothiocyanate, 5-isothiocyanatomethyl-1,3-benzodioxole, 3,4-methylenedioxybenzyl isothiocyanate, 5-isothiocyanatomethyl-2h-1,3-benzodioxole, 5-isothiocyanatomethyl-benzo 1,3 dioxole, 5-isothiocyanatomethyl benzo d 1,3 dioxole, acmc-20angn, pubchem9883, akos bbr-002798
C1OC2=C(O1)C=C(C=C2)CN=C=S
193.22
193.23

Chemical Identifiers

4430-47-1
193.22
PUJWRDBPAFJUJW-UHFFFAOYSA-N
2795371
C1OC2=C(O1)C=C(C=C2)CN=C=S
C9H7NO2S
MFCD00041217
3,4-methylenedioxy benzyl isothiocyanate, 1,3-benzodioxol-5-ylmethyl isothiocyanate, 5-isothiocyanatomethyl-1,3-benzodioxole, 3,4-methylenedioxybenzyl isothiocyanate, 5-isothiocyanatomethyl-2h-1,3-benzodioxole, 5-isothiocyanatomethyl-benzo 1,3 dioxole, 5-isothiocyanatomethyl benzo d 1,3 dioxole, acmc-20angn, pubchem9883, akos bbr-002798
5-(isothiocyanatomethyl)-1,3-benzodioxole
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

Documents