Learn More
3,4-(Methylenedioxy)aniline, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 171470500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3, 4-(Methylenedioxy)aniline | |
14268-66-7 | |
100.0 | |
Black to Brown | |
142°C | |
96% min. (GC) | |
C7H7NO2 | |
50 g | |
Solubility in water: slightly soluble | |
C1OC2=C(O1)C=C(C=C2)N | |
137.14 | |
137.14 | |
Low Melting Solid |
97% | |
96.0 | |
38.0°C to 43.0°C | |
144.0°C (16.0 mmHg) | |
Authentic | |
Glass bottle | |
MFCD00005832 | |
3,4-methylenedioxy aniline, benzo d 1,3 dioxol-5-amine, 5-amino-1,3-benzodioxole, 3,4-methylenedioxyaniline, 4-amino-1,2-methylenedioxybenzene, 5-aminobenzodioxole, methylenedioxyaniline, 2h-1,3-benzodioxol-5-amine, 1-amino-3,4-methylenedioxybenzene, 1-amino-3,4-dimethylenedioxybenzene | |
XGNXYCFREOZBOL-UHFFFAOYSA-N | |
1,3-benzodioxol-5-amine | |
84310 | |
97% |
Chemical Identifiers
14268-66-7 | |
137.14 | |
XGNXYCFREOZBOL-UHFFFAOYSA-N | |
84310 | |
C1OC2=C(O1)C=C(C=C2)N |
C7H7NO2 | |
MFCD00005832 | |
3,4-methylenedioxy aniline, benzo d 1,3 dioxol-5-amine, 5-amino-1,3-benzodioxole, 3,4-methylenedioxyaniline, 4-amino-1,2-methylenedioxybenzene, 5-aminobenzodioxole, methylenedioxyaniline, 2h-1,3-benzodioxol-5-amine, 1-amino-3,4-methylenedioxybenzene, 1-amino-3,4-dimethylenedioxybenzene | |
1,3-benzodioxol-5-amine |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
GHS Signal Word: Warning
EINECSNumber : 238-161-6
TSCA : TSCA
RUO – Research Use Only