Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(4-Methylbenzylidene)camphor, 99+%

$57.09 - $694.22

Chemical Identifiers

Chemical Name or Material 3-(4-Methylbenzylidene)camphor
CAS 36861-47-9
Molecular Formula C18H22O
Formula Weight 254.37
MDL Number MFCD00209662
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1656614

 
alfa aesar™
A1656614
25g Each for $57.09

AAA1656622

 
alfa aesar™
A1656622
100g Each for $167.78

AAA1656636

 
alfa aesar™
A1656636
500g Each for $694.22
Description & Specifications

Specifications

Formula Weight 254.37
Chemical Name or Material 3-(4-Methylbenzylidene)camphor
TSCA TSCA
Assay ≥99%
Molecular Formula C18H22O
MDL Number MFCD00209662
CAS 36861-47-9
Health Hazard 1 Warning
Melting Point 66° to 70°C
Quantity 25g
Health Hazard 2 GHS H Statement
H361
Suspected of damaging fertility or the unborn child.
Health Hazard 3 GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number DT5099565
Safety and Handling

Warning

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H361
Suspected of damaging fertility or the unborn child.