missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-[(4-Methylbenzyl)sulfonyl]-1H-1,2,4-triazole

$102.05 - $364.81

Chemical Identifiers

CAS 338393-13-8
Molecular Formula C10H11N3O2S
Formula Weight 237.28
MDL Number MFCD01568307
Chemical Name or Material 3-[(4-Methylbenzyl)sulfonyl]-1H-1,2,4-triazole
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5064903 SDS Alfa Aesar™
H5064903
1g Each for $364.81
AAH50649MD SDS Alfa Aesar™
H50649MD
250mg Each for $102.05
Specifications Description & Specifications

Chemical Identifiers

338393-13-8
237.28
3-[(4-Methylbenzyl)sulfonyl]-1H-1,2,4-triazole
C10H11N3O2S
MFCD01568307

Specifications

338393-13-8
237.28
1g
C10H11N3O2S
MFCD01568307
3-[(4-Methylbenzyl)sulfonyl]-1H-1,2,4-triazole

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning