Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(4-Methylbenzoyl)acrylic acid, 98%

$94.93 - $94.93

Chemical Identifiers

Chemical Name or Material 3-(4-Methylbenzoyl)acrylic acid
CAS 20972-36-5
Molecular Formula C11H10O3
Formula Weight 190.2
MDL Number MFCD00075338
Beilstein 2691231
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1055106

  SDS
alfa aesar™
L1055106
5g Each for $94.93
Description & Specifications

Specifications

Formula Weight 190.2
Chemical Name or Material 3-(4-Methylbenzoyl)acrylic acid
Assay 98%
MDL Number MFCD00075338
Molecular Formula C11H10O3
CAS 20972-36-5
Health Hazard 1 Warning
Quantity 5g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 2691231
Melting Point 138° to 140°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.