missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(4-Methylbenzoyl)acrylic acid, 98%

Manufacturer:  Alfa Aesar™ L1055106

 View more versions of this product

Catalog No. AAL1055106



Specifications

Specifications

3-(4-Methylbenzoyl)acrylic acid
5g
C11H10O3
2691231
VNJMEFZKYZHWEO-VOTSOKGWSA-N
(E)-4-(4-methylphenyl)-4-oxobut-2-enoic acid
691106
98%
138°C to 140°C
20972-36-5
MFCD00075338
3-4-methylbenzoyl acrylic acid, 2e-4-4-methylphenyl-4-oxobut-2-enoic acid, e-4-oxo-4-p-tolyl but-2-enoic acid, trans-3-4-methylbenzoyl acrylic acid, e-3-p-toluoylacrylic acid, chembl44664, regid_for_cid_691106, 3-4-methyl benzoyl acrylic acid, 3-4-methyl-benzoyl acrylic acid, 4-4-methylphenyl-4-oxo-2-butenoic acid
CC1=CC=C(C=C1)C(=O)C=CC(=O)O
190.198
190.2

Chemical Identifiers

20972-36-5
190.198
VNJMEFZKYZHWEO-VOTSOKGWSA-N
691106
CC1=CC=C(C=C1)C(=O)C=CC(=O)O
C11H10O3
MFCD00075338
3-4-methylbenzoyl acrylic acid, 2e-4-4-methylphenyl-4-oxobut-2-enoic acid, e-4-oxo-4-p-tolyl but-2-enoic acid, trans-3-4-methylbenzoyl acrylic acid, e-3-p-toluoylacrylic acid, chembl44664, regid_for_cid_691106, 3-4-methyl benzoyl acrylic acid, 3-4-methyl-benzoyl acrylic acid, 4-4-methylphenyl-4-oxo-2-butenoic acid
(E)-4-(4-methylphenyl)-4-oxobut-2-enoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents