missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-[(4-Methyl-1-piperidinyl)sulfonyl]benzeneboronic acid, 97%, Alfa Aesar™

$208.93 - $668.59

Chemical Identifiers

CAS 1449132-41-5
Molecular Formula C12H18BNO4S
Molecular Weight (g/mol) 283.149
MDL Number MFCD20265316
InChI Key KCFNPZMWUJEXKJ-UHFFFAOYSA-N
Synonym 3-4-methyl-1-piperidinyl sulfonyl benzeneboronic acid
PubChem CID 73995916
IUPAC Name [3-(4-methylpiperidin-1-yl)sulfonylphenyl]boronic acid
SMILES B(C1=CC(=CC=C1)S(=O)(=O)N2CCC(CC2)C)(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH54664MD View Documents Thermo Fisher Scientific
H54664MD
250mg Each for $208.93
 
AAH5466403 View Documents Thermo Fisher Scientific
H5466403
1g Each for $668.59
 
Specifications

Chemical Identifiers

1449132-41-5
283.149
KCFNPZMWUJEXKJ-UHFFFAOYSA-N
73995916
B(C1=CC(=CC=C1)S(=O)(=O)N2CCC(CC2)C)(O)O
C12H18BNO4S
MFCD20265316
3-4-methyl-1-piperidinyl sulfonyl benzeneboronic acid
[3-(4-methylpiperidin-1-yl)sulfonylphenyl]boronic acid

Specifications

250mg
C12H18BNO4S
3-4-methyl-1-piperidinyl sulfonyl benzeneboronic acid
B(C1=CC(=CC=C1)S(=O)(=O)N2CCC(CC2)C)(O)O
283.149
283.15
3-[(4-Methyl-1-piperidinyl)sulfonyl]benzeneboronic acid
1449132-41-5
MFCD20265316
KCFNPZMWUJEXKJ-UHFFFAOYSA-N
[3-(4-methylpiperidin-1-yl)sulfonylphenyl]boronic acid
73995916
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures