missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(4-Methoxyphenyl)propionic Acid 98.0+%, TCI America™

Manufacturer:  Tci America M103225G

 View more versions of this product

Catalog No. M103225G



Specifications

Specifications

3-(4-Methoxyphenyl)propionic Acid
Undesignated
25g
C10H12O3
3-4-methoxy-phenyl-propionic acid, 3-4-methoxyphenyl propanoic acid, 3-4-methoxyphenyl propionic acid, 3-4-methoxyphenyl-propionic acid, 3-p-methoxyphenyl propionic acid, 4-methoxyhydro cinnamic acid, 4-methoxyhydrocinnamic acid, benzenepropanoic acid, 4-methoxy, hydrocinnamic acid, p-methoxy, p-methoxyhydrocinnamic acid
COC1=CC=C(C=C1)CCC(=O)O
180.203
180.20
≥98.0% (GC,T)
194°C
104°C
1929-29-9
MFCD00002777
FIUFLISGGHNPSM-UHFFFAOYSA-N
3-(4-methoxyphenyl)propanoic acid
95750
Crystal-Powder at 20°C

Chemical Identifiers

1929-29-9
180.203
FIUFLISGGHNPSM-UHFFFAOYSA-N
95750
COC1=CC=C(C=C1)CCC(=O)O
C10H12O3
MFCD00002777
3-4-methoxy-phenyl-propionic acid, 3-4-methoxyphenyl propanoic acid, 3-4-methoxyphenyl propionic acid, 3-4-methoxyphenyl-propionic acid, 3-p-methoxyphenyl propionic acid, 4-methoxyhydro cinnamic acid, 4-methoxyhydrocinnamic acid, benzenepropanoic acid, 4-methoxy, hydrocinnamic acid, p-methoxy, p-methoxyhydrocinnamic acid
3-(4-methoxyphenyl)propanoic acid
Safety and Handling

Safety and Handling

TSCA : No

Documents