missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(4-Methoxyphenyl)propionic acid, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 126080250

 View more versions of this product

Catalog No. AC126080250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥97.5%
Crystalline Powder
White to light beige
100.0
p-Methoxyhydrocinnamic acid
C10H12O3
MFCD00002777
Glass bottle
Authentic
180.2
25g
99° to 103°C
3-(4-Methoxyphenyl)propionic acid
98%
1929-29-9
97.5
CH3OC6H4CH2CH2CO2H

Chemical Identifiers

1929-29-9
100.0
C10H12O3
180.2
p-Methoxyhydrocinnamic acid
≥97.5%
97.5
98%
CH3OC6H4CH2CH2CO2H
MFCD00002777
3-(4-Methoxyphenyl)propionic acid

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.