Please login to your online account to display your discounted pricing

3-(4-Methoxyphenyl)propionic acid, 98%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  126080100

 View more versions of this product

Catalog No. AC126080100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 180.2
Quantity 10g
Color White to light beige
Melting Point 99° to 103°C
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C10H12O3
CAS 1929-29-9
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 3-(4-Methoxyphenyl)propionic acid
Infrared Spectrum Authentic
Linear Formula CH3OC6H4CH2CH2CO2H
MDL Number MFCD00002777
Synonym p-Methoxyhydrocinnamic acid
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 3-(4-Methoxyphenyl)propionic acid
CAS 1929-29-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H12O3
Linear Formula CH3OC6H4CH2CH2CO2H
Formula Weight 180.2
MDL Number MFCD00002777
Synonym p-Methoxyhydrocinnamic acid
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents