Please login to your online account to display your discounted pricing

3-(4-Methoxyphenyl)propionic acid, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-(4-Methoxyphenyl)propionic acid
CAS 1929-29-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H12O3
Linear Formula CH3OC6H4CH2CH2CO2H
Formula Weight 180.2
MDL Number MFCD00002777
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC126080250

  SDS
thermo fisher scientific
126080250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 180.2
Physical Form Crystalline Powder
Quantity 25g
Color White to light beige
Melting Point 99° to 103°C
CAS Max % 100.0
Chemical Name or Material 3-(4-Methoxyphenyl)propionic acid
Synonym p-Methoxyhydrocinnamic acid
Assay Percent Range 98%
Molecular Formula C10H12O3
CAS 1929-29-9
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00002777
CAS Min % 97.5
Packaging Glass bottle
Linear Formula CH3OC6H4CH2CH2CO2H
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.