Please login to your online account to display your discounted pricing

3-(4-Methoxyphenyl)-5-methyl-4-isoxazolecarbonyl chloride, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 3-(4-Methoxyphenyl)-5-methyl-4-isoxazolecarbonyl chloride
CAS 465514-03-8
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H10ClNO3
Formula Weight 251.67
MDL Number MFCD03407359
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MO00394CB

  SDS
fisher scientific company
MO00394CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 251.67
Quantity 250mg
Assay 97%
CAS Min % 97.0
Chemical Name or Material 3-(4-Methoxyphenyl)-5-methyl-4-isoxazolecarbonyl chloride
CAS 465514-03-8
Health Hazard 1 GHS Signal Word: Danger
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C12H10ClNO3
MDL Number MFCD03407359
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.