Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(4-Methoxyphenyl)-2-phenylpropionic acid

$109.85 - $382.72

Chemical Identifiers

Chemical Name or Material 3-(4-Methoxyphenyl)-2-phenylpropionic acid
CAS 4314-68-5
Molecular Formula C16H16O3
Formula Weight 256.3
MDL Number MFCD00046532
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5021303

  SDS
alfa aesar™
H5021303
1g Each for $382.72

AAH50213MD

  SDS
alfa aesar™
H50213MD
250mg Each for $109.85
Description & Specifications

Specifications

Formula Weight 256.3
Chemical Name or Material 3-(4-Methoxyphenyl)-2-phenylpropionic acid
Molecular Formula C16H16O3
MDL Number MFCD00046532
CAS 4314-68-5
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.