Please login to your online account to display your discounted pricing

3-(4-Methoxyphenoxy)benzaldehyde, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-(4-Methoxyphenoxy)benzaldehyde
Name Note 95%
CAS 62373-80-2
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C14 H12 O3
Linear Formula CH3OC6H4OC6H4CHO
Formula Weight 228.25
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC187700100

  SDS
thermo fisher scientific
187700100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Formula Weight 228.25
Physical Form Liquid
Quantity 10g
Color Clear yellow
TSCA TSCA
Density 1.0800g/mL
EINECS Number 263-527-7
CAS Max % 100.0
CAS Min % 94.0
Refractive Index 1.5920 to 1.5970 (20°C, 589nm)
Packaging Glass bottle
Infrared Spectrum Authentic
Name Note 95%
Chemical Name or Material 3-(4-Methoxyphenoxy)benzaldehyde
Molecular Formula C14 H12 O3
CAS 62373-80-2
Linear Formula CH3OC6H4OC6H4CHO
MDL Number MFCD00003358
Assay Percent Range 95%