missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(4-Methoxybenzylideneamino)benzeneboronic acid pinacol ester

$36.64 - $568.00

Chemical Identifiers

CAS 380151-91-7
Molecular Formula C20H24BNO3
Formula Weight 337.22
MDL Number MFCD11113043
Synonym N-(4-Methoxybenzylidene)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Chemical Name or Material 3-(4-Methoxybenzylideneamino)benzeneboronic acid pinacol ester
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5112903 SDS Alfa Aesar™
H5112903
1g Each for $36.64
AAH5112906 SDS Alfa Aesar™
H5112906
5g Each for $139.76
AAH5112914 SDS Alfa Aesar™
H5112914
25g Each for $568.00
Specifications Description & Specifications

Chemical Identifiers

380151-91-7
337.22
N-(4-Methoxybenzylidene)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
C20H24BNO3
MFCD11113043
3-(4-Methoxybenzylideneamino)benzeneboronic acid pinacol ester

Specifications

1g
C20H24BNO3
MFCD11113043
3-(4-Methoxybenzylideneamino)benzeneboronic acid pinacol ester
380151-91-7
337.22
N-(4-Methoxybenzylidene)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline