missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(4-Methoxybenzoyl)propionic Acid 98.0+%, TCI America™

$83.94 - $261.69

Chemical Identifiers

CAS 3153-44-4
Molecular Formula C11H12O4
Molecular Weight (g/mol) 208.213
MDL Number MFCD00002795
InChI Key OMTDIBZSUZNVJK-UHFFFAOYSA-N
Synonym 3-4-methoxybenzoyl propanoic acid
PubChem CID 76613
IUPAC Name 4-(4-methoxyphenyl)-4-oxobutanoic acid
SMILES COC1=CC=C(C=C1)C(=O)CCC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M13835G View Documents Tci America
M13835G
5g Each for $83.94
 
M138325G View Documents Tci America
M138325G
25g Each for $261.69
 
Specifications

Chemical Identifiers

3153-44-4
208.213
OMTDIBZSUZNVJK-UHFFFAOYSA-N
76613
COC1=CC=C(C=C1)C(=O)CCC(=O)O
C11H12O4
MFCD00002795
3-4-methoxybenzoyl propanoic acid
4-(4-methoxyphenyl)-4-oxobutanoic acid

Specifications

White-Yellow
5g
C11H12O4
3-4-methoxybenzoyl propanoic acid
COC1=CC=C(C=C1)C(=O)CCC(=O)O
208.213
208.21
≥98.0% (HPLC,T)
149°C
3153-44-4
MFCD00002795
OMTDIBZSUZNVJK-UHFFFAOYSA-N
4-(4-methoxyphenyl)-4-oxobutanoic acid
76613
Crystal-Powder at 20°C
3-(4-Methoxybenzoyl)propionic Acid
Safety and Handling

Safety and Handling

TSCA : No