Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(4-Methoxybenzoyl)propionic acid, 98+%

$574.08 - $574.08

Chemical Identifiers

Chemical Name or Material 3-(4-Methoxybenzoyl)propionic acid
CAS 3153-44-4
Molecular Formula C11H12O4
Formula Weight 208.21
MDL Number MFCD00002795
Beilstein 785074
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1821022

  SDS
alfa aesar™
A1821022
100g Each for $574.08

AAA1821006

  SDS
alfa aesar™
A1821006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1821014

  SDS
alfa aesar™
A1821014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 3-(4-Methoxybenzoyl)propionic acid
Melting Point 147° to 151°C
Quantity 100g
CAS 3153-44-4
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay ≥98%
Molecular Formula C11H12O4
Formula Weight 208.21
MDL Number MFCD00002795
Beilstein 785074
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.