missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(4-Methoxybenzoyl)propionic Acid 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 125750250

 View more versions of this product

Catalog No. AC125750250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥96.0%
208.21
White
10, 958
96.0
97%
MFCD00002795
3-(4-Methoxybenzoyl)propionic acid
3153-44-4
Crystalline Powder
25g
148° to 150°C
100.0
Authentic
C11H12O4
97%
CH3OC6H4COCH2CH2CO2H

Chemical Identifiers

3153-44-4
100.0
C11H12O4
208.21
3-(4-Methoxybenzoyl)propionic acid, 97%
96.0
97%
CH3OC6H4COCH2CH2CO2H
MFCD00002795
≥96.0%

Safety and Handling

SDS & Safety Documents

EINECSNumber : 221-593-4

TSCA : TSCA