Please login to your online account to display your discounted pricing

3-(4-Methoxybenzoyl)propionic Acid 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-(4-Methoxybenzoyl)propionic acid
Name Note 97%
CAS 3153-44-4
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C11H12O4
Linear Formula CH3OC6H4COCH2CH2CO2H
Formula Weight 208.21
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC125750250

  SDS
thermo fisher scientific
125750250
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥96.0%
Formula Weight 208.21
Quantity 25g
Color White
EINECS Number 221-593-4
TSCA TSCA
Melting Point 148° to 150°C
Beilstein 10, 958
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 3-(4-Methoxybenzoyl)propionic acid
Infrared Spectrum Authentic
Name Note 97%
Assay Percent Range 97%
Molecular Formula C11H12O4
Linear Formula CH3OC6H4COCH2CH2CO2H
MDL Number MFCD00002795
CAS 3153-44-4