Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-[4-Methoxy-3-(trifluoromethyl)phenyl]propionic acid, 97%

$79.73 - $264.76

Chemical Identifiers

Chemical Name or Material 3-[4-Methoxy-3-(trifluoromethyl)phenyl]propionic acid
CAS 1005205-20-8
Molecular Formula C11H11F3O3
Formula Weight 248.2
MDL Number MFCD11519317
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3242803

 
alfa aesar™
H3242803
1g Each for $264.76

AAH32428MD

 
alfa aesar™
H32428MD
250mg Each for $79.73
Description & Specifications

Specifications

Formula Weight 248.2
Chemical Name or Material 3-[4-Methoxy-3-(trifluoromethyl)phenyl]propionic acid
Assay 97%
Molecular Formula C11H11F3O3
CAS 1005205-20-8
Melting Point 94° to 97°C
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
MDL Number MFCD11519317
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.