Please login to your online account to display your discounted pricing

3,4-Lutidine 98%, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  125205000

 View more versions of this product

Catalog No. AC125205000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay 97.5 to 102.5% Total Base, ≥97.5% (GC)
Physical Form Liquid
Formula Weight 107.15
Color Clear yellow to brownish
Quantity 500g
TSCA TSCA
RTECS Number OK9800000
EINECS Number 209-511-5
Density 0.9500g/mL
Beilstein 20, 246
CAS Max % 100.0
Infrared Spectrum Authentic
Chemical Name or Material 3, 4-Lutidine
Packaging Glass bottle
Refractive Index 1.5100 to 1.5130 (20°C, 589nm)
Assay Percent Range 98%
CAS 583-58-4
Synonym 3, 4-Dimethylpyridine
CAS Min % 97.5
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Toxic in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 98%
Molecular Formula C7H9N
MDL Number MFCD00006403

Specifications

Chemical Name or Material 3, 4-Lutidine
Name Note 98%
CAS 583-58-4
CAS Min % 97.5
CAS Max % 100.0
Assay 97.5 to 102.5% Total Base, ≥97.5% (GC)
Assay Percent Range 98%
Molecular Formula C7H9N
Formula Weight 107.15
MDL Number MFCD00006403
Synonym 3, 4-Dimethylpyridine
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Toxic in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Flammable liquid and vapor.

SDS & Safety Documents