Please login to your online account to display your discounted pricing

3,4-Lutidine 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3, 4-Lutidine
Name Note 98%
CAS 583-58-4
CAS Min % 97.5
CAS Max % 100.0
Assay 97.5 to 102.5% total base, 97.5% min. (GC)
Assay Percent Range 98%
Molecular Formula C7H9N
Formula Weight 107.15g/mol
MDL Number MFCD00006403
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC125205000

  SDS
thermo fisher scientific
125205000
Glass bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.5100 to 1.5130 (20°C, 589nm)
Density 0.9500g/mL
Color Clear yellow to brown
Quantity 500g
Chemical Name or Material 3, 4-Lutidine
Name Note 98%
CAS 583-58-4
CAS Min % 97.5
CAS Max % 100.0
Physical Form Clear yellow to brownish liquid
Assay 97.5 to 102.5% total base, 97.5% min. (GC)
Assay Percent Range 98%
Molecular Formula C7H9N
Formula Weight 107.15g/mol
MDL Number MFCD00006403
Beilstein 20, 246
Synonym 3, 4-Dimethylpyridine
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Toxic in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RTECS Number OK9800000
EINECS Number 209-511-5
TSCA TSCA
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Toxic in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Flammable liquid and vapor.