Please login to your online account to display your discounted pricing

3-(4-hydroxyphenyl)propanohydrazide, Maybridge


Quantity 10g

Manufacturer: fisher scientific company  BTB00937EA

 View more versions of this product

Catalog No. BTB00937EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 180.21
Quantity 10g
Assay 97%
Chemical Name or Material 3-(4-hydroxyphenyl)propanohydrazide
Molecular Formula C9H12N2O2
CAS 65330-63-4
Health Hazard 1 GHS Signal Word: Warning

Specifications

Chemical Name or Material 3-(4-hydroxyphenyl)propanohydrazide
Assay 97%
Molecular Formula C9H12N2O2
Formula Weight 180.21
Accessories

GHS Signal Word: Warning