missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Hexanedione, TCI America™

$105.42 - $316.25

Chemical Identifiers

CAS 4437-51-8
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00010237
InChI Key KVFQMAZOBTXCAZ-UHFFFAOYSA-N
Synonym 3,4-hexanedione, bipropionyl, dipropionyl, diethyl diketone, 3,4-hexane dione, unii-4m4npr5vgd, 3,4-hexandione, fema no. 3168, ccris 6295, 4m4npr5vgd
PubChem CID 62539
IUPAC Name hexane-3,4-dione
SMILES CCC(=O)C(=O)CC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H05735ML View Documents TCI America
H05735ML
5mL Each for $105.42
 
H057325ML View Documents TCI America
H057325ML
25mL Each for $316.25
 
Specifications

Chemical Identifiers

4437-51-8
114.144
KVFQMAZOBTXCAZ-UHFFFAOYSA-N
62539
CCC(=O)C(=O)CC
C6H10O2
MFCD00010237
3,4-hexanedione, bipropionyl, dipropionyl, diethyl diketone, 3,4-hexane dione, unii-4m4npr5vgd, 3,4-hexandione, fema no. 3168, ccris 6295, 4m4npr5vgd
hexane-3,4-dione

Specifications

130°C
5mL
C6H10O2
1224
KVFQMAZOBTXCAZ-UHFFFAOYSA-N
hexane-3,4-dione
62539
Clear Liquid at 20°C
3,4-Hexanedione
Yellow
4437-51-8
MFCD00010237
3,4-hexanedione, bipropionyl, dipropionyl, diethyl diketone, 3,4-hexane dione, unii-4m4npr5vgd, 3,4-hexandione, fema no. 3168, ccris 6295, 4m4npr5vgd
CCC(=O)C(=O)CC
114.144
114.14
≥96.0% (GC)
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Freezer