Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Hexanedione, 95%

Chemical Identifiers

Chemical Name or Material 3,4-Hexanedione
CAS 4437-51-8
Molecular Formula C6H10O2
Formula Weight 114.15
MDL Number MFCD00010237
UN Number UN1224
Beilstein 1700837
Synonym Dipropionyl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0665514

  SDS
alfa aesar™
L0665514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0665522

  SDS
alfa aesar™
L0665522
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 114.15
Chemical Name or Material 3,4-Hexanedione
MDL Number MFCD00010237
CAS 4437-51-8
UN Number UN1224
Synonym Dipropionyl
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
Quantity 25g
TSCA TSCA
Assay 95%
Molecular Formula C6H10O2
Beilstein 1700837
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapor.
Flash Point 29°C (84°F)
Boiling Point 129° to 131°C
Melting Point −10°C
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapor.