Please login to your online account to display your discounted pricing

3-(4-Fluorophenyl)propionic acid, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-(4-Fluorophenyl)propionic acid
CAS 459-31-4
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H9FO2
Linear Formula COOH(CH2)2C6H4F
Formula Weight 168.17
MDL Number MFCD00060327
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC375610050

  SDS
thermo fisher scientific
AC375610050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 168.17
Physical Form Crystalline Powder or Crystals
Quantity 5g
Color White to off-white
Melting Point 86° to 91°C
CAS Max % 100.0
Linear Formula COOH(CH2)2C6H4F
Molecular Formula C9H9FO2
Assay Percent Range 98%
CAS 459-31-4
Chemical Name or Material 3-(4-Fluorophenyl)propionic acid
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00060327
CAS Min % 97.5
Packaging Glass bottle
Infrared Spectrum Authentic