Please login to your online account to display your discounted pricing

3-(4-Fluorophenyl)-5-methyl-4-isoxazolecarboxylic acid, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  MO00391DA

 View more versions of this product

Catalog No. MO00391DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 221.19
Quantity 1g
Assay 97%
CAS Min % 97.0
CAS 1736-21-6
Chemical Name or Material 3-(4-Fluorophenyl)-5-methyl-4-isoxazolecarboxylic acid
Assay Percent Range 97%
Molecular Formula C11H8FNO3
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
MDL Number MFCD03407356
Packaging Amber Glass Bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 3-(4-Fluorophenyl)-5-methyl-4-isoxazolecarboxylic acid
CAS 1736-21-6
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H8FNO3
Formula Weight 221.19
MDL Number MFCD03407356
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

SDS & Safety Documents