Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(4-Fluorophenyl)-4-methyl-1H-pyrazole

$929.68 - $929.68

Chemical Identifiers

Chemical Name or Material 3-(4-Fluorophenyl)-4-methyl-1H-pyrazole
Molecular Formula C10H9FN2
Formula Weight 176.2
MDL Number MFCD01940436
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5044003

  SDS
alfa aesar™
H5044003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5044006

  SDS
alfa aesar™
H5044006
5g Each for $929.68
Description & Specifications

Specifications

Formula Weight 176.2
Molecular Formula C10H9FN2
Chemical Name or Material 3-(4-Fluorophenyl)-4-methyl-1H-pyrazole
Quantity 1g
MDL Number MFCD01940436
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.