Please login to your online account to display your discounted pricing

3-(4-Fluorophenyl)-1H-pyrazole-4-carbaldehyde, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 3-(4-Fluorophenyl)-1H-pyrazole-4-carbaldehyde
CAS 306936-57-2
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H7FN2O
Formula Weight 190.18
MDL Number MFCD01922131
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SP01479DA

  SDS
fisher scientific company
SP01479DA
Amber glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 190.18
Quantity 1g
Assay 97%
CAS Min % 97.0
Assay Percent Range 97%
Chemical Name or Material 3-(4-Fluorophenyl)-1H-pyrazole-4-carbaldehyde
Molecular Formula C10H7FN2O
MDL Number MFCD01922131
CAS 306936-57-2
Packaging Amber glass bottle