Please login to your online account to display your discounted pricing

3-(4-Fluorophenyl)-1H-pyrazole-4-carbaldehyde, 97%, Maybridge


Quantity 1g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  SP01479DA

 View more versions of this product

Catalog No. SP01479DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 190.18
Quantity 1g
Assay 97%
CAS Min % 97.0
Assay Percent Range 97%
Chemical Name or Material 3-(4-Fluorophenyl)-1H-pyrazole-4-carbaldehyde
Molecular Formula C10H7FN2O
MDL Number MFCD01922131
CAS 306936-57-2
Packaging Amber glass bottle

Specifications

Chemical Name or Material 3-(4-Fluorophenyl)-1H-pyrazole-4-carbaldehyde
CAS 306936-57-2
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H7FN2O
Formula Weight 190.18
MDL Number MFCD01922131