missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(4-Fluorophenyl)-1-isopropylindole, 98%

$34.24 - $112.58

Chemical Identifiers

CAS 93957-49-4
Molecular Formula C17H16FN
Molecular Weight (g/mol) 253.32
MDL Number MFCD01075733
InChI Key ZDZJOIIBECYKAJ-UHFFFAOYSA-N
Synonym 3-4-fluorophenyl-1-isopropyl-1h-indole, 1-isopropyl-3-4-fluorophenyl indole, 3-4-fluorophenyl-1-isopropylindole, 3-4-fluorophenyl-1-1-methylethyl-1h-indole, 1-isopropyl-3-4-flrorophenyl-indole, 1h-indole, 3-4-fluorophenyl-1-1-methylethyl, 4-fluorophenyl-1-isopropyl-3-indole, 3-4-fluorophenyl-1-propan-2-ylindole, 3-4-fluoro-phenyl-1-isopropyl-1h-indole, 3-4-fluorophenyl-1-methylethyl indole
PubChem CID 736229
IUPAC Name 3-(4-fluorophenyl)-1-propan-2-ylindole
SMILES CC(C)N1C=C(C2=CC=CC=C21)C3=CC=C(C=C3)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1634103 View Documents Alfa Aesar™
L1634103
1g Each for $34.24
 
AAL1634106 View Documents Alfa Aesar™
L1634106
5g Each for $112.58
 
Specifications

Chemical Identifiers

93957-49-4
253.32
ZDZJOIIBECYKAJ-UHFFFAOYSA-N
736229
CC(C)N1C=C(C2=CC=CC=C21)C3=CC=C(C=C3)F
C17H16FN
MFCD01075733
3-4-fluorophenyl-1-isopropyl-1h-indole, 1-isopropyl-3-4-fluorophenyl indole, 3-4-fluorophenyl-1-isopropylindole, 3-4-fluorophenyl-1-1-methylethyl-1h-indole, 1-isopropyl-3-4-flrorophenyl-indole, 1h-indole, 3-4-fluorophenyl-1-1-methylethyl, 4-fluorophenyl-1-isopropyl-3-indole, 3-4-fluorophenyl-1-propan-2-ylindole, 3-4-fluoro-phenyl-1-isopropyl-1h-indole, 3-4-fluorophenyl-1-methylethyl indole
3-(4-fluorophenyl)-1-propan-2-ylindole

Specifications

95°C to 98°C
93957-49-4
MFCD01075733
3-4-fluorophenyl-1-isopropyl-1h-indole, 1-isopropyl-3-4-fluorophenyl indole, 3-4-fluorophenyl-1-isopropylindole, 3-4-fluorophenyl-1-1-methylethyl-1h-indole, 1-isopropyl-3-4-flrorophenyl-indole, 1h-indole, 3-4-fluorophenyl-1-1-methylethyl, 4-fluorophenyl-1-isopropyl-3-indole, 3-4-fluorophenyl-1-propan-2-ylindole, 3-4-fluoro-phenyl-1-isopropyl-1h-indole, 3-4-fluorophenyl-1-methylethyl indole
CC(C)N1C=C(C2=CC=CC=C21)C3=CC=C(C=C3)F
253.32
253.32
3-(4-Fluorophenyl)-1-isopropylindole
1g
C17H16FN
4457093
ZDZJOIIBECYKAJ-UHFFFAOYSA-N
3-(4-fluorophenyl)-1-propan-2-ylindole
736229
98%